Learn More
Epredia™ Histoplast Paraffin
Shop All Epredia Products
Click to view available options
Product Type:
Paraffin IM
Paraffin Kit PE
Paraffin LP
Paraffin PE
Unit Size:
Pack of 2
Pack of 8
Description
- Made of paraffin and plastic polymers
- Delivers excellent processing and yields thin, wrinkle-free sections
Ordering Information:
Packaged in convenient, easy-to-open, resealable 2 lb. bags. Also available in a premeasured paraffin kit for use on the Epredia™ Excelsior™ AS Tissue Processor (waste drawer included).
Specifications
Specifications
| For Use With (Equipment) | Epredia, Excelsior AS, and Revos Tissue Processors |
| Product Type | Paraffin Kit PE |
| Quantity | 2 x 2.3 kg (Plus Waste Drawer) |
| For Use With (Application) | Can be used in both infiltration and embedding procedures |