Learn More
Cytiva His Mag Sepharose Excel (10%)
Shop All Cytiva ProductsDescription
His Mag Sepharose Excel are magnetic beads designed for rapid, small-scale purification and screening of his-tagged proteins consisting of an Immobilized Metal Affinity Chromatography (IMAC) resin precharged with nickel ions that are strongly bound to a chelating ligand.
- Simplify small-scale purification and expression screening of histidine-tagged proteins secreted in eukaryotic cell culture
- Robust parallel screening with high reproducibility
- Increase target protein yield and decrease degradation through reduced and simplified sample handling
- Capture histidine-tagged protein from different sample volumes (low microliter to high milliliter scale)
- Visible and dense Sepharose based magnetic beads give increased simplicity for handling
- High density of beads ensures rapid capture by magnetic devices
Small-scale capture, purification, and screening of histidine-tagged proteins that are expressed and secreted into supernatants from eukaryotic cells such as insect cells or Chinese Hamster Ovary (CHO) cells in HyClone HyCell CHO cell culture media.
The base of the matrix is based on Sepharose resin, incorporated with magnetite.
Compatible with MagRack 6 and MagRack Maxi, separation tools for handling the beads in microcentrifuge tubes.
Specifications
Specifications
| Binding Capacity | < At least 10 mg (histidine)6-tagged protein/ml sedimented medium |
| Matrix | Highly cross-linked spherical agarose including magnetite |
| Content And Storage | 4°C to 30°C, 20% ethanol, 10% medium slurry |
| Quantity | 100 mL |
| For Use With (Application) | Immunoprecipitation and purification of biotinulated biomolecules |
| Ligand Type | IMAC immobilized with Ni |
Safety and Handling
- His Mag Sepharose excel, 100 ml (10%)
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol, nickel
Your input is important to us. Please complete this form to provide feedback related to the content on this product.