missing translation for 'onlineSavingsMsg'
Learn More

Wesbart™ Hinged Cryobox Rack

Wesbart™ Hinged Cryobox Rack supplied with trays to hold 100-cell hinged cryoboxes, suitable for upright freezers

Brand:  Wesbart™ BH304/4NMT

Product Code. 16262461

  • € 192.00 / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Aluminum front access racks
  • Supplied with trays specially designed to hold 100 cell hinged cryoboxes
  • For use with upright freezers
  • Racks are suitable for cryoboxes with dimensions of 141 x 151 x 63 mm
Specifications

Specifications

Cryobox Rack
Aluminum
Upright Freezers
Cryoboxes (12, 63 x 141 x 151 mm)
4 Trays/Rack
560 x 150 x 287 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.