Learn More
Cytiva HiLoad™ Superdex™ 200 PG XK Column
Product Code. 11369991 Shop All Cytiva Products
Click to view available options
Bed Volume:
180mL
480mL
1000mL
Unit Size:
Each
Product Code. 11369991
Brand: Cytiva 90100040
Prepacked XK columns designed for high-resolution preparative gel filtration chromatography.
HiLoad™ Superdex™ 200 prep grade are prepacked XK columns designed for high-resolution preparative gel filtration chromatography. The column is an excellent choice for separating MAbs from dimers and contaminants.
  • High resolution with short run times and good recovery.
  • High capacity at high flow rates.
  • Separations can be scaled up to production levels.
  • Prepacked for convenience and reproducibility.
  • 1/16” fittings for convenient connections to ÄKTA design systems.
Superdex™ prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these resins are determined by the dextran component. Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr < 10 000 (Superdex™ 30 pg), Mr 3 000 to 70 000 (Superdex™ 75 pg), and Mr 10 000 600 000 (Superdex™ 200 pg). The resin combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions.
TRUSTED_SUSTAINABILITY
Specifications
Bed Volume 1000mL
Product Type HiLoad Superdex
Material Borosilicate Glass Column
Content And Storage 4 to 30°C, 20% Ethanol
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.