Learn More
Nabertherm™ High-Temperature Ovens, Forced Convection Chamber Furnaces
Chamber furnaces with air circulation are characterized by their extremely high temperature uniformity. Suitable for processes such as annealing, crystalizing, preheating, curing, and tool making.
€ 7395.00 - € 30630.00
Specifications
Temperature (Metric) 850°C
View More Specs
Products 10
Product Code Brand Capacity (Metric) Controller Type Price Quantity & Availability  
Product Code Brand Capacity (Metric) Controller Type Price Quantity & Availability  
17338049
View Documents
Nabertherm™
NA--0683K2
60 L B500
€ 12110.00
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17348049
View Documents
Nabertherm™
NA--0683M2
60 L C540
€ 12435.00
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17398049
View Documents
Nabertherm™
NA--1283K2
120 L B500
€ 15765.00
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17308059
View Documents
Nabertherm™
NA--1283M2
120 L C540
€ 16095.00
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17358059
View Documents
Nabertherm™
NA--2583K2
250 L B400
€ 21855.00
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
17368059
View Documents
Nabertherm™
NA--2583M2
250 L C540
€ 22185.00
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
17318069
View Documents
Nabertherm™
NA--5083K2
500 L B400
€ 30280.00
Each
Estimated Shipment: 02-07-2025
to see stock.
Add to basket
17328069
View Documents
Nabertherm™
NA--5083M2
500 L C540
€ 30630.00
Each
Estimated Shipment: 02-07-2025
to see stock.
Add to basket
17378069
View Documents
Nabertherm™
NAT-0381L1
30 L B510 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17388069
View Documents
Nabertherm™
NAT-0381O1
30 L C550 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Horizontal air circulation with optimum distribution through stainless steel baffles
  • Swing door hinged on the right
  • Temperature uniformity up to +/- 4 C according to DIN 17052-1 (NAT 15/65 and NAT 30/85 up to +/- 5°C) in the empty work space
  • One frame sheet and rails for two additional trays included in the scope of delivery (NAT 15/65 and NAT 30/85 without frame sheet)
  • Base frame included in the delivery, NAT 15/65 and NAT 30/85 designed as table-top model
Specifications
Specifications
850°C
Documents
Product Certifications
Product Content Correction
Product Title
Nabertherm™ High-Temperature Ovens, Forced Convection Chamber Furnaces