Learn More
Aalborg™ High Resolution Valve Stainless Steel
Aalborg™ High Resolution Valve Stainless Steel is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
Brand: Aalborg™ VM4-SV-2A
This item is not returnable.
View return policy
Description
- For high resolution
- Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
- They may also be used as shut off valves.
- Bodies of the valves are reinforced by structurally rigid metallic shells.
- Bushings are made of plated brass or 316 stainless steel.
Specifications
2500 mL/min. | |
121°C | |
70mL/min. | |
High Resolution Valve |
Product Content Correction
Product Title
Aalborg™ High Resolution Valve Stainless Steel > Air Flow Rate: 2500mL/min.