Learn More
Aalborg™ High Resolution Valve Stainless Steel
Aalborg™ High Resolution Valve Stainless Steel is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
Brand:  Aalborg™ VM4-SV-2A
 View more versions of this product
Product Code. 11787549
  • € 249.00 / Each
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For high resolution
  • Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
  • They may also be used as shut off valves.
  • Bodies of the valves are reinforced by structurally rigid metallic shells.
  • Bushings are made of plated brass or 316 stainless steel.
Specifications
Specifications
2500 mL/min.
121°C
70mL/min.
High Resolution Valve
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Aalborg™ High Resolution Valve Stainless Steel > Air Flow Rate: 2500mL/min.