Learn More
Heidolph Instruments High Hood XL
For use as an add on to incubating shakers
Brand:  Heidolph Instruments 549-90060-00
1254.45 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 10792014
  • € 1515.00 / Each
Estimated Shipment: 16-06-2025
to see stock.
Explore more special offers
Description
With a height of 428 mm, this cap is particularly suitable for tall culture vessels, such as 2000 mL Erlenmeyer flasks
TRUSTED_SUSTAINABILITY
Specifications
Specifications
115/230 V
5°C to 65°C
428 mm
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Heidolph Instruments High Hood XL > High Hood XL; Height: 428 mm