Learn More
Thermo Scientific Chemicals High Form Nickel Crucible;Cap (ml), 50;Top Dia (mm), 44;Bottom Dia (mm), 32;Depth (mm), 51
Product Code. 15493575
Click to view available options
Capacity (Metric):
20 mL
30 mL
50 mL
75 mL
100 mL
250 mL
500 mL
Diameter (Metric) Outer:
38mm
41mm
44mm
54mm
61mm
84mm
101mm
Height (Metric):
35mm
43mm
51mm
57mm
62mm
78mm
91mm
Unit Size:
Each
Product Code. 15493575
Brand: Thermo Scientific Chemicals 035883.KT
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description High Form
Material Nickel
Capacity (Metric) 20 mL
Diameter (Metric) Outer 38mm
Height (Metric) 35mm
Product Type Nickel Crucible
Diameter (Metric) Base 22mm