Learn More
Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories
Product Code. 11851571 Shop All Thermo Scientific Products
Click to view available options
Product Type:
Combustible Gas Connection
Cover
General Accessories
Lead Through, 10 to 13 mm ID
Lead Through, 3 mm ID
Non-combustible Gas Connection
Side Window Modifications, Left Hand
Side Window Modifications, Right Hand
Solenoid Valves
Vacuum Tap Connection
Water Tap Connection
Wind Screen Protection Kit
Work Surface Wraps
For Use With (Equipment):
Heraguard ECO 0.9
Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95; Herasafe 2030i 0.9, 1.2, 1.5, 1.8; Maxisafe 2030i 0.9, 1.2, 1.5, 1.8
Heraguard ECO 1.2
Heraguard ECO 1.2, 1.5,1.8, 1.8/95
Heraguard ECO 1.5
Heraguard ECO 1.8
Heraguard ECO 1.8/95
Unit Size:
Each
Product Code. 11851571
Brand: Thermo Scientific™ 50076408
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations.
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations. Options include media valves, solenoid valve, stainless-steel work surface wraps, side window modifications, UV and UV night covers, windscreen protection kits and additional accessories.
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
Description Lead through for hose connection for side wall use. Suitable for 3mm inner diameter hose
Product Type Lead Through, 3 mm ID
Configuration Options Field Installed