missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemoglobin, freeze dried, >98%, MP Biomedicals™
Chemical Identifiers
CAS | 9008-02-0 |
---|---|
Molecular Formula | C13H10N2O2 |
Molecular Weight (g/mol) | 226.24 |
MDL Number | MFCD00131282 |
InChI Key | INGWEZCOABYORO-UHFFFAOYSA-N |
Synonym | 7-methyl-2-2-furyl-1,8-naphthyridine-4 1h-one, d0y8up, 2-furan-2-yl-7-methyl-1h-1,8 naphthyridin-4-one |
PubChem CID | 13285535 |
IUPAC Name | 2-(furan-2-yl)-7-methyl-1H-1,8-naphthyridin-4-one |
SMILES | CC1=CC=C2C(=O)C=C(NC2=N1)C1=CC=CO1 |
Chemical Identifiers
9008-02-0 | |
226.24 | |
INGWEZCOABYORO-UHFFFAOYSA-N | |
13285535 | |
CC1=CC=C2C(=O)C=C(NC2=N1)C1=CC=CO1 |
C13H10N2O2 | |
MFCD00131282 | |
7-methyl-2-2-furyl-1,8-naphthyridine-4 1h-one, d0y8up, 2-furan-2-yl-7-methyl-1h-1,8 naphthyridin-4-one | |
2-(furan-2-yl)-7-methyl-1H-1,8-naphthyridin-4-one |
Specifications
9008-02-0 | |
>98% | |
MFCD00131282 | |
INGWEZCOABYORO-UHFFFAOYSA-N | |
2-(furan-2-yl)-7-methyl-1H-1,8-naphthyridin-4-one | |
13285535 | |
Crystalline Powder |
Brown-Red | |
C13H10N2O2 | |
7-methyl-2-2-furyl-1,8-naphthyridine-4 1h-one, d0y8up, 2-furan-2-yl-7-methyl-1h-1,8 naphthyridin-4-one | |
CC1=CC=C2C(=O)C=C(NC2=N1)C1=CC=CO1 | |
226.24 | |
≈64,500 | |
Hemoglobin |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hemoglobin, freeze dried, >98%, MP Biomedicals™