Learn More
Helios Italquartz Quartz Crucibles
Click to view available options
Capacity (Metric):
15 mL
20 mL
25 mL
30 mL
50 mL
75 mL
130 mL
190 mL
Diameter (Metric) Outer:
35mm
41mm
43mm
47mm
50mm
57mm
67mm
75mm
80mm
Color:
Translucent
Transparent
Description:
Short form
Tall form
Unit Size:
Each
Description
- Chemically inert to acids (except HF)
- Available in a wide range of volumes
- Available in transparent or translucent color
- Available in tall or short form
- Resistant to corrosion
Specifications
Specifications
| Color | Transparent |
| Description | Tall form |
| Material | Quartz |
| Max. Temperature (Metric) | 1000°C |
| Capacity (Metric) | 20 mL |
| Diameter (Metric) Outer | 35mm |
| Height (Metric) | 38mm |
| Product Type | Crucible |