Learn More
Heidolph Instruments AUTOaccurate-Sensor
Sensor Automatic Distillation
Brand: Heidolph Instruments 569-00040-00
Description
- For Hei-VAP Expert Control and Hei-VAP Ultimate Control
- Only in combination with glassware set G3 and G6
Order Info
dimType
Specifications
Aluminum | |
Sensor Automatic Distillation | |
1 Ea. | |
115/230 VAC |
Product Content Correction
Product Title
Heidolph Instruments AUTOaccurate-Sensor > Type: Sensor Automatic Distillation