Learn More
Heidolph™ Hei-FLOW Silver 2 Package
This package includes Hei-FLOW Value 06, pump head SP standard 2.5, 1 m each Tygon and silicone tubing.
Brand: Heidolph™ 523-50068-00
2810.98 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
This package includes:
- Hei-FLOW Value 06
- Pump head SP standard 2.5
- 1 m each Tygon™ (standard) and Silicone tubing (inner Ø 6.4 mm)
Specifications
EU Plug | |
LED | |
230 V |
Digital Variable Speed | |
IP33 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Heidolph™ Hei-FLOW Silver 2 Package > EU Plug