missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Heclin
HECT E3 ubiquitin ligase inhibitor
€ 204.00 - € 771.00
Specifications
Purity | >98% |
---|---|
Product Type | Heclin |
Molecular Weight (g/mol) | 283.32 |
Inhibitors | Heclin |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780108
|
Tocris Bioscience™
5433/10 |
10 mg |
€ 204.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18704529
|
Tocris Bioscience™
5433/50 |
50 mg |
€ 771.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HECT E3 ubiquitin ligase inhibitor; inhibits Nedd4, Smurf2 and WWP1 (IC50 values are 6.3, 6.8 and 6.9 μM respectively). Selective for HECT-mediated over RING-mediated ubiquitination.Specifications
>98% | |
Heclin | |
283.32 | |
Heclin |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title