Learn More
3M™ Heavy Duty EPDM Breathing Tube, SS-BT-55
3M™ Heavy Duty EPDM Breathing Tube, SS-BT-55 is a fixed-length EPDM rubber breathing tube designed for use with approved powered air systems.
Brand: 3M™ 7100236678
This item is not returnable.
View return policy
Description
- Fixed-length breathing tube made from EPDM rubber
- Approved for use with 3M™ Powered Air Respirator PF-600E
- Suitable for use with specific 3M™ Promask™ Full Facepiece Reusable Respirator, FM3 and 3M™ Vision™ Full Facepiece Reusable Respirator with 3M™ Powered Air Respirator PF-600E
- Protective breathing tube covers available to lengthen service life of breathing tubes
Specifications
EPDM Rubber | |
Breathing Tube | |
3M™ PAPR Systems | |
SS-BT-55 |