Learn More
Bibby Scientific™ Electrothermal™ Heating Element
Product Code. p-8011213
Click to view available options
Capacity (Metric):
100 mL
500 mL
Unit Size:
Each
Product Code. 11517497
Brand: Bibby Scientific™ 1008661
Only null left
Add to basket
Add to basket
Spare heating elements for Soxhlet Air Baths. Bibby Scientific™ Electrothermal™ Heating Elements are available for 100mL and 500mL flask capacities.
  • Spare heating elements for Soxhlet Air Baths.
  • Available in 100mL and 500mL capacities
  • Spare heating elements for Soxhlet Air Baths.
  • Available in 100mL and 500mL capacities
Specifications
Capacity (Metric) 100 mL
For Use With Spare Soxhlet Extractor
Product Type Heating Element