Learn More
Horst™ Heating Cable, Quartz Yarn, High Temperature Resistance
High temperatures, small winding radii, high power rate.
Brand: Horst™ 21002
This item is not returnable.
View return policy
Description
- Flexible.
- High temperature resistance.
- High power rates per meter.
- Quartz yarn insulation.
- Small winding radii.
Specifications
Bending radius: 10 mm, Thickness: 5 mm | |
230 V Nominal | |
1.5 m | |
3.5 to 4.5 mm |
370 m | |
Quartz Yarn | |
2.1 W | |
900°C (Nominal) |