Learn More
Eppendorf™ Heat Sealer S100
Shop All Eppendorf Products
Click to view available options
Plug Type:
EU Plug
Unit Size:
Each
Description
- Hermetic sealing of multiwell plates
- Fixed sealing temperature for reproducible results
- Eliminates evaporation in PCR, reducing cross contamination
- Seals 96- and 384-well plates
- Compact design, portable device
- Integrated thermostats prevents overheating
- Improved mechanics for even easier sealing
- PCR plate adapter included
Specifications
Specifications
| Hertz | 50/60 Hz |
| Voltage | 100/230 V |
| Dimensions (L x W x H) | 220 × 250 × 230 mm |
| Electrical Requirements | 100/230 V, 50/60 Hz |
| For Use With (Equipment) | 96- and 384-well plates |
| Plug Type | EU Plug |