Learn More
SCAT HDPE Safety Funnels With Hinged Lid
Shop All SCAT Europe Products
Click to view available options
Includes:
With hinged lid
Unit Size:
Each
Description
The SCAT safety funnel with hinged lid is designed to save space and is easy to handle. Due to the hinged lid as a closing mechanism, even very small quantities and viscous liquid waste can be discharged easily. Der Klappdeckel kann nach der Nutzung geschlossen werden, was Ausdunstungen Die Verschraubungen sind frei drehbar - dies erleichtert das Aufschrauben des Trichters. Alle Trichter sind aus schwarzem PE-HD, elektrisch leitfähig und mit einem Erdungskabel versehen, zur sicheren Erdung des Trichters. The dirt sieve serves as additional splash protection.
Specifications
Specifications
| Capacity (Metric) | 330 mL |
| Material | HDPE |
| Height (Metric) | 240 mm |
| Color | Red |
| Description | Thread size: S 60/61;;en; |
| Includes | With hinged lid |
| Quantity | 1 Ea. |
| Product Type | Safety Funnel |