Learn More
Kautex™ HDPE Natural Rectangular Narrow Neck Bottle
Brand: Kautex™ 2000070521
This item is not returnable.
View return policy
Description
- High stability, ribbed sides for secure grip
- Rectangular, natural color
- Caps are not included; sold separately.
Specifications
0.25 L | |
19 mm | |
HDPE | |
No | |
Square | |
Narrow Neck Bottle |
Natural | |
Storage, Packaging, Dispensing, Mailing | |
Narrow | |
Narrow | |
148 mm |