Learn More
VITLAB™ HDPE Measuring Scoops
Click to view available options
Capacity (Metric):
25 mL
Unit Size:
Pack of 12
Specifications
Specifications
| Capacity (Metric) | 25 mL |
| Color | Natural |
| Product Type | Measuring Scoop |
| Material | HDPE |
| Length (Metric) | 135 mm |