Learn More
Buerkle™ Hexagonal Nut
Product Code. 11991471 Shop All Bürkle Products
Click to view available options
Includes:
Inner Thread 12.7 mm
Inner Thread 19.05 mm
Inner Thread 25.4 mm
Inner Thread 50.8 mm
Unit Size:
Each
Product Code. 11991471
Brand: Buerkle™ 85370200
Only null left
Add to basket
Add to basket
Buerkle™ Hexagonal Nut with various sizes of inner threads.
  • Incudes inner thread
  • Available in black and white
  • Constructed of HDPE or PP
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Description Hexagonal shape
For Use With General Pump
Includes Inner Thread 50.8 mm
Material Polypropylene
Quantity 1 Ea.
Type Hex Nut