Learn More
Horst™ HBST Model Heating Tape
Click to view available options
Length (Metric):
1 m
10 m
2 m
3 m
4 m
5 m
Unit Size:
Each
Specifications
Specifications
| Dimensions (L x W) | 2 Lm |
| Dimensions (L x W x H) | 2 Lm |
| Length (Metric) | 2 m |
| Electrical Requirements | ∼230V Voltage |
| Model | HBST |
| Product Type | Heating Tape |