Learn More
Horst™ HBST Model Heating Tape
Brand:  Horst™ 20703
 View more versions of this product
Product Code. 10590392
  • € 216.00 / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Specifications
3 Lm
3 m
HBST
3 Lm
∼230V Voltage
Heating Tape
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Horst™ HBST Model Heating Tape > Length: 3m