Learn More
Horst™ HBQ Model Very Flexible Heating Tape
Brand:  Horst™ 20401
 View more versions of this product
Product Code. 10332322
  • € 317.00 / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Specifications
0.5 Lm
0.5 m
HBQ
0.5 Lm
∼230V Voltage
Very Flexible Heating Tape
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Horst™ HBQ Model Very Flexible Heating Tape > Length: 0.5m