Learn More
Horst™ HBQ Model Very Flexible Heating Tape
Click to view available options
Length (Metric):
0.5 m
1 m
1.5 m
2 m
3 m
Unit Size:
Each
Specifications
Specifications
| Dimensions (L x W) | 1 Lm |
| Dimensions (L x W x H) | 1 Lm |
| Length (Metric) | 1 m |
| Electrical Requirements | ∼230V Voltage |
| Model | HBQ |
| Product Type | Very Flexible Heating Tape |