Learn More
New Pig™ HazMat Spill Kits
Click to view available options
Capacity (Metric):
13.62 L
34.068 L
79.49 L
For Use With (Application):
Acids, Bases, Unknowns
Acids, Bases, Unknowns Fluid Absorption
Oils, Coolants, Solvents, Water
Unit Size:
Each
Description
- Feature PIG absorbents proven for rapid response
- Socks stop spreading spills; PIG HazMat Pads absorb quickly
- Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
- Temporary disposal bags help make cleanup easier
- Bright yellow bag and large, color-coded label make the kit easy to locate
- Strap for easy carrying and hanging
- Compact and easy to store
Specifications
Specifications
| Capacity (Metric) | 34.068 L |
| For Use With (Application) | Oils, Coolants, Solvents, Water |