Learn More
Hanna Instruments™ pH Electrode Cleaning Solution
Eliminates impurities and residues that are left on electrode surfaces when immersed in samples during measurement or when stored incorrectly. Hanna Instruments™ pH Electrode Cleaning Solution dissolves any mineral deposits or other coatings by soaking up to 20 minutes.
Brand:  Hanna Instruments™ HI7061L
Product Code. 10688301
  • € 42.55 / 460mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
General purpose cleaning solution that can be used for a variety of applications.
  • Eliminates impurities and residues that are left on electrode surfaces when immersed in samples during measurement or when stored incorrectly.
  • Dissolves any mineral deposits or other coatings by soaking up to 20 minutes.
Specifications
Specifications
500 mL
Aqueous
Removing impurities and residues from ph Electrodes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers