Learn More
Corning™ Hank's Balanced Salt Solutions
Click to view available options
Quantity:
1 x 500 mL
6 x 500 mL
Concentration:
1 X
With Additives:
Calcium, Magnesium
Unit Size:
Pack of 6
Without Additives:
No Calcium, No Magnesium
No Calcium, No Magnesium, No Phenol Red
Description
pH of 10X solutions must be adjusted following dilution to 1X concentration.
Specifications
Specifications
| Color | Orange Red |
| Content And Storage | 2° to 30°C |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | Hanks' Balanced Salt Solution |
| With Additives | Calcium, Magnesium |
| pH | 7.1 to 7.4 |
| Quantity | 1 x 500 mL |