Learn More
Cole-Parmer™ Hand Operated Vacuum Pumps
Produce up to 25 in Hg vacuum or test for pressure leaks with this vacuum pump. Cole-Parmer™ Hand Operated Vacuum Pumps are sealed and self-lubricated for a long, maintenance-free life.
Brand: Cole-Parmer™ 79301-01
163.51 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- Built-in trigger enables you to release vacuum without disconnecting the pump
- Zinc alloy is perfect for rugged applications; PVC is lightweight and easy to handle
Specifications
190.5 x 50.8 x 241.3 mm | |
Zinc Alloy | |
15 mL/stroke | |
Hand Operated Vacuum Pump |
Product Content Correction
Product Title
Cole-Parmer™ Hand Operated Vacuum Pumps > Capacity: 15mL/stroke