missing translation for 'onlineSavingsMsg'
Learn More

Hammond™ Desiccant, Cole-Parmer™

Product Code. p-8004049 Shop All Cole-Parmer Products
Click to view available options
Chemical Name or Material:
Indicating Drierite Desiccant
Non-Indicating Drierite Desiccant
Quantity:
1 lb.
5 lb.
Unit Size:
Pack of 12
Pack of 4
This item is not returnable. View return policy

Product Code. 11795609

Brand: Cole Parmer 0719305

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Choose indicating or non-indicating desiccant—indicating desiccant starts blue, then turns pink when regeneration or renewal is necessary. Use Hammond™; Drierite™ Desiccant (anhydrous calcium sulfate) in desiccators, drying columns or drying tubes.

  • Indicating desiccant drying capacity is 10 to 14% of its weight for water and gases 
  • Non-indicating desiccant drying capacity is 6% of its weight for water and gases

Specifications

Material Calcium Sulfate
Quantity 1 lb.
Suitability Used in Desiccators, Drying Columns, or Drying Tubes
Chemical Name or Material Indicating Drierite Desiccant
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Drierite, Indicating
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H400-Very toxic to aquatic life.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.