Learn More
IKA™ HABITAT Ferment, Double-walled 1 L Package
The HABITAT ferment package for the bioreactor HABITAT research contains a 1 L double-walled glass vessel for fermentation applications as well as the appropriate stirring drive.
Brand: IKA™ 0010007657
This item is not returnable.
View return policy
Description
Together with the separately available control tower package (e.g. HABITAT ferment) you receive all necessary components for a successful cultivation. For temperature control via the double jacket we recommend one of our our circulators such as the HRC 2 basic or control.
Scope of delivery
- Stand 1
- HA.gv.dw.1 Glass vessel, double-wall
- HA.mt.s.2 Motor, small
- HA.hp.s.1 Harvest pipe, straight
- HA.ino Port for inoculation
- HA.ba.1 Baffle
- HA.sp.r.1 Ring sparger, 0.5 mm
- HA.cn Condenser
- HA.ip.b.1 6-blade segment impeller
- HA.s.tm.1 Temperature sensor
- HA.s.ph.1 pH sensor
- HA.s.do.1 DO sensor
- HA.s.fo Foam sensor
- HA.s.lv.1 Level sensor
- HA.cab.dw Cable and tube set
- HA.sf.250 Sample flask (4 pcs.)
Specifications
Bioreactor Package | |
1 L | |
8.09 kg |
Autoclavable | |
Borosilicate Glass |
Product Content Correction
Product Title
IKA™ HABITAT Ferment, Double-walled 1 L Package > Double-walled, 1 L