Learn More
Thermo Scientific™ H-Flex 1 Swinging Bucket Rotor and Accessories
Shop All Thermo Scientific Products
Click to view available options
Description:
18 x 15 mL Adapter
33 x 10/12 mL Adapter
400 mL Bottle/Centrilab Adapter
42 x 5/7 mL Adapter
8 x 50 mL Adapter
Biocontainment Lids
Buckets
Cross
Microplate Adapter
Replacement O-Rings
Unit Size:
Each
Pack of 2
Description
Spin more with less! Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint General Purpose centrifuge. High capacity and flexibility—all in a small footprint.
- Auto-Lock and Click Seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
- Spins blood tubes, conical tubes and microplates without changing rotor or buckets
- Get results faster without the need to change hardware
- Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
Specifications
Specifications
| Description | Replacement O-Rings |
| Material | EPDM |
| Type | Swinging Bucket Rotor |
| For Use With (Equipment) | Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series |
| Biocontainment | Yes |
| Autoclavable | Not Autoclavable |
| Quantity | Set of 2 |