Learn More
Thermo Scientific™ H-Flex 1 Swinging Bucket Rotor and Accessories
Product Code. 16545311 Shop All Thermo Scientific Products
Click to view available options
Description:
18 x 15 mL Adapter
33 x 10/12 mL Adapter
400 mL Bottle/Centrilab Adapter
42 x 5/7 mL Adapter
8 x 50 mL Adapter
Biocontainment Lids
Buckets
Cross
Microplate Adapter
Replacement O-Rings
Unit Size:
Each
Pack of 2
Product Code. 16545311
Brand: Thermo Scientific™ 75003305
Spin more with less! High capacity and flexibility—all in a small footprint.
Spin more with less! Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint General Purpose centrifuge. High capacity and flexibility—all in a small footprint.
  • Auto-Lock and Click Seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
  • Spins blood tubes, conical tubes and microplates without changing rotor or buckets
  • Get results faster without the need to change hardware
  • Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
TRUSTED_SUSTAINABILITY
Specifications
Description 33 x 10/12 mL Adapter
Material Molded Polyamide Nylone
Type Swinging Bucket Rotor
Capacity 33 x 10/12 mL Blood Tubes Each Adapter
Max. Speed 4297
For Use With (Equipment) 75003300 H-Flex Rotor Cross and 75003301 Buckets
Biocontainment Yes
Autoclavable Autoclavable
Quantity Set of 2