Learn More
Kgw Isotherm™ GSS Stainless Steel Dewar Flasks
Click to view available options
Capacity (Metric):
500 mL
1 L
6 L
Height (Metric):
156 mm
180 mm
206 mm
270 mm
Unit Size:
Each
Specifications
Specifications
| For Use With (Application) | Used to Store and Transport liquid nitrogen ( LN2 ), Dry Ice ( CO2 ) and Other Coolants |
| Height (Metric) | 270 mm |
| Includes | Loose Lid Insulating Plug |
| Capacity (Metric) | 6 L |
| Diameter (Metric) Inner | 184 mm |
| Material | GSS Stainless Steel |