Learn More
Kgw Isotherm™ GSS Stainless Steel Dewar Flasks
Brand: Kgw Isotherm™ 2305
Specifications
Used to Store and Transport liquid nitrogen ( LN2 ), Dry Ice ( CO2 ) and Other Coolants | |
Loose Lid Insulating Plug | |
184 mm |
270 mm | |
6 L | |
GSS Stainless Steel |