Learn More
GSK0660, MedChemExpress
MedChemExpress GSK0660 is a potent antagonist of PPARβ and PPARδ, with IC50s of 155 nM for both isoforms.
Brand:  Medchem Express HY-12377
Encompass_Preferred
 View more versions of this product
Product Code. 16467847
  • € 120.15 / 5mg
Explore more special offers
Description
Description
GSK0660 is a potent antagonist of PPARβ and PPARδ, with IC50s of 155 nM for both isoforms.
Specifications
Specifications
GSK0660
Light Yellow
C19H18N2O5S2
DMSO : ≥ 49 mg/mL (117.09 mM)
418.49
99.55%
Solid
1014691-61-2
Research
5 mg
O=C(C1=C(S(=O)(NC2=CC=C(NC3=CC=CC=C3)C=C2OC)=O)C=CS1)OC
418.49
Research
Metabolism-protein/nucleotide metabolism
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only