Learn More
Tocris Bioscience™ GSK 2837808A
Potent and selective LDHA inhibitor
Brand:  Tocris Bioscience™ 5189/10
322.24 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18767058
  • € 416.00 / 10mg
Explore more special offers
Description
Description
GSK 2837808A is a potent and selective lactate dehydrogenase A and B (LDHA and LDHB) inhibitor (IC50 values are 2.6 and 43 nM for LDHA and LDHB respectively). Inhibits lactate production in selected cancer cell lines. Reduces glucose uptake and enhances mitochondrial oxygen consumption in Snu398 hepatocellular carcinoma cells. Inhibits proliferation and induces apoptosis in Snu398 cells. Inhibits transcription of histone 2B (H2B) gene in HCT116 and NCM460 cells. Cell permeable.
Specifications
Specifications
98%
1445879-21-9
10 mg
M.W. (Observed): 649.62
3-[[3-[(Cyclopropylamino)sulfonyl]-7-(2,4-dimethoxy-5-pyrimidinyl)-4-quinolinyl]amino]-5-(3,5-difluorophenoxy)benzoic acid
-20°C
GSK 2837808A
GSK 2837808A
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only