missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Growth Hormone Releasing Hexapeptide

€ 126.00 - € 299.00

Specifications

CAS 87616-84-0
Content And Storage Store at -20°C
Format Lyophilized powder
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15447709
View Documents Promotion Details
Thermo Scientific™
J66373.#0
5 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15457709
View Documents Promotion Details
Thermo Scientific™
J66373.MB
25 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

87616-84-0
Store at -20°C
Lyophilized powder
Videos
SDS
Documents

Documents

Product Certifications