Learn More
Karl Hecht™ Ground Edge Microscope Slides with Frosted End
Product Code. p-7147870
Click to view available options
Frosted:
Both Ends
Unit Size:
Pack of 50
Product Code. 15904030
Brand: Karl Hecht™ 42407010
Only null left
Add to basket
Add to basket
Selected glass quality microscope slides which have been pre-cleaned and are ready for use.
  • Available with one or two frosted ends
  • Half-white
  • CE marked
  • Non-corrosive
  • Ground edges 90°
  • Standard thickness of approx. 1mm
  • Size approx. 76 x 26mm
  • 50/box, 5000/carton
Specifications
Length (Metric) 76 mm
Width (Metric) 26 mm
No. per Case 5000/Carton
No. per Pack 50/Box
Edges 90°, Ground
Frosted Both Ends
Material Glass
Precleaned Yes
Thickness (Metric) 1 mm
Color Transparent
Certifications/Compliance CE
For Use With (Application) Microscopy, staining
Dimensions (L x W) 76 x 26 mm