Learn More
Notrax Gripper Sof-Tred™ Dyna-Shield™ Layer with Lengthwise Corrugated Rib Pattern
Notrax Gripper Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.
Brand:  Notrax 413S0023BY
 View more versions of this product
Product Code. 17742098
  • € 56.00 / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Description
  • Lengthwise corrugated rib pattern for sure footing
  • Exceptional worker comfort due to 127 mm thick specially formulated compound
  • Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
  • Available with yellow borders along sides comply with OSHA code 1910-144
  • All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications
Specifications
Black/Yellow
91 cm
Industrial and Commercial Matting
36 in.
Microcellular Vinyl
60 cm