Learn More
Liebherr™ Grid Shelves for Refrigerator
Product Code. p-8002219 Shop All Liebherr Products
Click to view available options
For Use With (Equipment):
Model LKPv14 65
Unit Size:
Each
Product Code. 12110132
Brand: Liebherr™ 7112393
Only null left
Add to basket
Add to basket
Use these grid shelves with freezers for general laboratory storage of temperature-sensitive samples.
  • Grid shelf for use with Pharmacy refrigerators and Laboratory refrigerators with Comfort electronic controllers
  • Grid shelf size: 510 x 650 mm
Specifications
Type Grid Shelf
Shelves Size: 510 x 650 mm
Cabinet Material Stainless Steel
For Use With (Equipment) Model LKPv14 65
Model LKPv 1423, LKPv 1420, LKPv 6520, LKPv 6523, LGPv 1420, LGPv 6520
Depth (Metric) Exterior 650 mm
Width (Metric) Exterior 510 mm