Learn More
Greiner Bio-One Speedy Quick Release Tube Holder
Trigger mechanism for safe and easy needle ejection
Brand:  Greiner Bio-One 450212
Product Code. 10748315
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Speedy Quick Release Tube Holder
  • After venipuncture the needle is simply released from the holder by pushing the release mechanism so that the needle falls directly into a sharps disposal container
  • The release mechanism is designed to retract automatically
  • It is recommended to use this device as a single-use holder
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tube Holder
Non-sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers