Learn More
Greiner Bio-One Blood Collection Tubes
Can be used for CSF determinations
Brand: Greiner Bio-One 455001
This item is not returnable.
View return policy
Description
Can be used for CSF determinations
Polyethylene Therephthalate
Specifications
Evacuated Blood Collection Tube | |
Non-ridged Pull Cap | |
White/Black | |
Blood collection |
No Additive | |
9 mL | |
16 × 100 mm |