Learn More
Greiner Bio-One Serum Tubes: Clot Activator with Gel Separator
For routine clinical chemistry, immunohematology, serology or immunology testing
Marca: Greiner Bio-One 456018
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Coated with micronized silica particles which activate clotting when tubes are gently inverted
- Tubes with gel contain a barrier gel on the tube bottom that during centrifugation moves upward to the serum-clot interface to form a stable barrier, separating serum from cells
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- Tubes have an 18-month shelf life from the date of manufacture
- Pediatric tubes come with white rings for quick identification
- Safety Twist Caps can be removed and replaced several times
- Nonscrew caps can be removed and replaced as often as required
Compliance
Tubes with gel separator are FDA 510(k) cleared for therapeutic drug monitoring (TDM)
Especificaciones
Microscopic Silica Particles with Separation Gel | |
Non-ridged Pull Cap | |
Gold |
Plastic | |
5 mL |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido