Learn More
Witeg™ Graduated Erlenmeyer Flasks With Screw Cap
Witeg™ Graduated Erlenmeyer Flasks With Screw Cap are graduated and designed for storage of culture media and mixing.
Brand:  Witeg™ 5 507 008
 View more versions of this product
Product Code. 10431551
  • € 108.00 / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Description
  • Borosilicate glass 3.3 construction
  • Graduated
  • Screw cap included
  • Available in a variety of volume capacities
Specifications
Specifications
3000 mL
Screw cap
187 mm
310 mm
Screw Cap