Learn More
Witeg™ Graduated Erlenmeyer Flasks With Screw Cap
Witeg™ Graduated Erlenmeyer Flasks With Screw Cap are graduated and designed for storage of culture media and mixing.
Brand:  Witeg™ 5 507 004
 View more versions of this product
Product Code. 10238480
  • € 242.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Borosilicate glass 3.3 construction
  • Graduated
  • Screw cap included
  • Available in a variety of volume capacities
Specifications
Specifications
250 mL
Screw cap
85 mm
140 mm
Screw Cap