Learn More
Fisherbrand™ Grade 605 Very Absorbent Joseph Paper
Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
120 x 120 mm
150 x 150 mm
220 x 400 mm
500 x 350 mm
Format:
Folded Sheet
Sheet
Packaging Quantity:
100 Pk.
200 Pk.
50 Pk.
500 Pk.
Unit Size:
Pack of 100
Pack of 200
Pack of 50
Pack of 500
Description
Applications:
- Cleaning and drying of, glass containers, tubes, flasks, microscopic plates, bottles
Specifications
Specifications
| Length (Metric) | 500 mm |
| Width (Metric) | 350 mm |
| Dimensions (L x W) | 500 x 350 mm |
| Format | Folded Sheet |
| For Use With (Application) | Cleaning and drying of, Glass containers, tubes, flasks, microscopic plates, bottles in laboratories and hospitals |
| Packaging Quantity | 50 Pk. |
| Grade | 605 |