Learn More
Thermo Scientific™ Grade 25 Ruby Spheres, Dia.: 2mm
Click to view available options
Quantity:
100 Pieces
25 Pieces
5 Pieces
Unit Size:
Pack of 100
Pack of 25
Pack of 5
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Product Type | Ruby Sphere |
| Diameter (Metric) | 2 mm |
| Grade | Grade 25 |
| Quantity | 100 Pieces |