Learn More
Grace™ Reveleris™ SRC C18 Cartridges
Ideal for speed, resolution and capacity
€ 177.53
Specifications
Material | C18 |
---|---|
For Use With (Application) | For Speed, Resolution and Capacity, Conventional AppliCatIons, Polar Compounds and Popular Flash Systems |
Max. Pressure | 200 psi |
Product Code | Brand | Bed Weight (Metric) | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight (Metric) | Type | Price | Quantity & Availability | ||||||
11333581
|
Grace™
145152103 |
12 g | Reveleris SRC C18 Cartridge | N/A | N/A | N/A | |||||
Description
- Withstand higher pressures (up to 200psig) and deliver greater loading capacity
- Superior, high load silica for excellent resolution
- Optimally packed to give high plate count
- Tighter particle size distribution and minimal fines keep backpressure low
- RFID tag streamlines Reveleris system (sold separately) operation by auto-loading cartridge specifications
- Compatible with other popular flash systems
Specifications
C18 | |
For Speed, Resolution and Capacity, Conventional AppliCatIons, Polar Compounds and Popular Flash Systems | |
200 psi |
Product Title
Grace™ Reveleris™ SRC C18 Cartridges